Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G57300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085291.3 | NP_001078760.1 | 2 - 3 | 270 | 139 | |
NM_001203629.1 | NP_001190558.1 | 2 - 3 | 263 | 139 | ||
NM_001345255.1 | NP_001332423.1 | 1 - 2 | 142 | 139 | ||
NM_125112.5 | NP_200540.1 | 2 - 3 | 220 | 139 | ||
GenBank mRNA | AY050939.1 | - | 2 - 3 | 219 | 139 | |
AY091173.1 | - | 1 - 2 | 93 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |