Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203630.1 | NP_001190559.1 | 14 - 15 | 2434 | 111 | |
NM_001345258.1 | NP_001331203.1 | 10 - 11 | 2213 | 111 | ||
NM_001345259.1 | NP_001331204.1 | 14 - 15 | 2488 | 111 | ||
NM_125118.3 | NP_200545.1 | 14 - 15 | 2550 | 111 | ||
GenBank mRNA | AY072153.1 | - | 14 - 15 | 2430 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |