Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203630.1 | NP_001190559.1 | 3 - 4 | 208 | 88 | |
NM_001345259.1 | NP_001331204.1 | 3 - 4 | 262 | 88 | ||
NM_125118.3 | NP_200545.1 | 3 - 4 | 324 | 88 | ||
GenBank mRNA | AY072153.1 | - | 3 - 4 | 204 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |