Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XBAT32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125157.3 | NP_200582.3 | 5 - 6 | 1158 | 81 | |
GenBank mRNA | BT011688.1 | - | 5 - 6 | 1030 | 81 | |
BT012272.1 | - | 5 - 6 | 653 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |