Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085295.2 | NP_001078764.1 | 6 - 7 | 908 | 102 | |
NM_001345284.1 | NP_001332019.1 | 5 - 6 | 869 | 102 | ||
NM_001345285.1 | NP_001332020.1 | 5 - 6 | 981 | 102 | ||
GenBank mRNA | DQ503426.1 | - | 5 - 6 | 777 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |