Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125208.3 | NP_200631.1 | 4 - 5 | 1046 | 102 | |
GenBank mRNA | AF016846.1 | - | 4 - 5 | 1021 | 102 | |
AY059874.1 | - | 4 - 5 | 1026 | 102 | ||
AY114646.1 | - | 4 - 5 | 946 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |