Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125246.4 | NP_200667.2 | 2 - 3 | 523 | 137 | |
GenBank mRNA | BT024924.1 | - | 2 - 3 | 302 | 137 | |
U62742.2 | - | 2 - 3 | 347 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |