Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345328.1 | NP_001330402.1 | 3 - 4 | 776 | 71 | |
NM_001345329.1 | NP_001330401.1 | 3 - 4 | 503 | 71 | ||
NM_125281.1 | NP_200699.1 | 2 - 3 | 374 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |