Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125372.4 | NP_200788.1 | 1 - 2 | 259 | 100 | |
GenBank mRNA | BT014975.1 | - | 1 - 2 | 178 | 100 | |
BT015848.1 | - | 1 - 2 | 149 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |