Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203644.1 | NP_001190573.1 | 19 - 20 | 3066 | 133 | |
NM_125397.3 | NP_200812.2 | 19 - 20 | 2911 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |