Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037029.1 | NP_001032106.1 | 3 - 4 | 574 | 69 | |
NM_001085305.1 | NP_001078774.1 | 3 - 4 | 574 | 69 | ||
NM_125429.4 | NP_568921.1 | 3 - 4 | 811 | 69 | ||
GenBank mRNA | AF083703.1 | - | 3 - 4 | 492 | 69 | |
AF233883.1 | - | 3 - 4 | 436 | 69 | ||
AF360273.1 | - | 3 - 4 | 466 | 69 | ||
AK317029.1 | - | 3 - 4 | 499 | 69 | ||
AK317496.1 | - | 3 - 4 | 499 | 69 | ||
AY088662.1 | - | 3 - 4 | 506 | 69 | ||
BT000673.1 | - | 3 - 4 | 499 | 69 | ||
BT000676.1 | - | 3 - 4 | 495 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |