Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP2;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125459.4 | NP_200874.1 | 2 - 3 | 731 | 72 | |
GenBank mRNA | AK228720.1 | - | 2 - 3 | 514 | 72 | |
AY087245.1 | - | 2 - 3 | 514 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |