Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125462.4 | NP_200877.1 | 10 - 11 | 1948 | 73 | |
GenBank mRNA | AF188994.1 | - | 10 - 11 | 1699 | 73 | |
AF233592.1 | - | 10 - 11 | 1382 | 73 | ||
AK229564.1 | - | 10 - 11 | 1906 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |