Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345457.1 | NP_001332102.1 | 4 - 5 | 1228 | 63 | |
NM_125514.5 | NP_200929.1 | 4 - 5 | 1059 | 63 | ||
GenBank mRNA | AY057627.1 | - | 4 - 5 | 974 | 63 | |
AY141994.1 | - | 3 - 4 | 704 | 63 | ||
X92419.1 | - | 945 | 63 | |||
X92420.1 | - | 890 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |