Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345475.1 | NP_001330106.1 | 1 - 2 | 394 | 87 | |
NM_125537.3 | NP_200952.1 | 1 - 2 | 394 | 87 | ||
GenBank mRNA | AF144389.1 | - | 1 - 2 | 286 | 87 | |
AK229043.1 | - | 1 - 2 | 339 | 87 | ||
BT024467.1 | - | 1 - 2 | 181 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |