Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125578.3 | NP_568941.1 | 1 - 2 | 541 | 65 | |
GenBank mRNA | AY054180.1 | - | 1 - 2 | 332 | 65 | |
AY066039.1 | - | 1 - 2 | 101 | 65 | ||
KJ138631.1 | - | 1 - 2 | 101 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |