Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345543.1 | NP_001318863.1 | 2 - 3 | 155 | 77 | |
NM_125619.2 | NP_201031.1 | 1 - 2 | 178 | 77 | ||
GenBank mRNA | DQ056733.1 | - | 1 - 2 | 48 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |