Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125633.5 | NP_201045.1 | 2 - 3 | 1402 | 87 | |
GenBank mRNA | AY093078.1 | - | 2 - 3 | 1172 | 87 | |
AY128766.1 | - | 2 - 3 | 1128 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |