Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103841.3 | NP_175376.1 | 1 | 563 | 68 | |
GenBank mRNA | BT011230.1 | - | 1 | 563 | 68 | |
BT012158.1 | - | 1 | 463 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |