Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP47A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333392.1 | NP_001321910.1 | 2 - 3 | 1187 | 66 | |
NM_001333393.1 | NP_001321909.1 | 2 - 3 | 1208 | 66 | ||
NM_103848.3 | NP_175383.1 | 2 - 3 | 1208 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |