Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198253.1 | NP_001185182.1 | 1 - 2 | 145 | 130 | |
NM_103855.5 | NP_175390.2 | 1 - 2 | 183 | 130 | ||
GenBank mRNA | AY062728.1 | - | 1 - 2 | 139 | 130 | |
AY072306.1 | - | 1 - 2 | 144 | 130 | ||
AY093342.1 | - | 1 - 2 | 58 | 130 | ||
BT008393.1 | - | 1 - 2 | 58 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |