Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198253.1 | NP_001185182.1 | 9 - 10 | 1154 | 59 | |
NM_103855.5 | NP_175390.2 | 9 - 10 | 1192 | 59 | ||
GenBank mRNA | AY062728.1 | - | 9 - 10 | 1148 | 59 | |
AY072306.1 | - | 9 - 10 | 1153 | 59 | ||
AY093342.1 | - | 9 - 10 | 1067 | 59 | ||
BT008393.1 | - | 9 - 10 | 1067 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |