Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103858.5 | NP_175393.1 | 3 - 4 | 1056 | 154 | |
GenBank mRNA | AJ345085.1 | - | 3 - 4 | 795 | 154 | |
AY054522.1 | - | 3 - 4 | 954 | 154 | ||
BT002570.1 | - | 3 - 4 | 795 | 154 | ||
KJ138969.1 | - | 3 - 4 | 795 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |