Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198255.1 | NP_001185184.1 | 8 - 9 | 2040 | 59 | |
NM_103863.3 | NP_564554.1 | 8 - 9 | 2039 | 59 | ||
GenBank mRNA | AF370517.1 | - | 8 - 9 | 2034 | 59 | |
AY064622.1 | - | 8 - 9 | 1774 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |