Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333424.1 | NP_001323380.1 | 3 - 4 | 440 | 142 | |
NM_001333425.1 | NP_001323379.1 | 2 - 3 | 532 | 142 | ||
NM_103902.4 | NP_564562.1 | 3 - 4 | 450 | 142 | ||
GenBank mRNA | BT004060.1 | - | 3 - 4 | 377 | 142 | |
BT005145.1 | - | 3 - 4 | 361 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |