Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198259.1 | NP_001185188.1 | 20 - 21 | 2073 | 113 | |
NM_001333440.1 | NP_001322424.1 | 19 - 20 | 2303 | 113 | ||
NM_001333441.1 | NP_001322422.1 | 20 - 21 | 2090 | 113 | ||
NM_001333442.1 | NP_001322423.1 | 20 - 21 | 2058 | 113 | ||
NM_103933.6 | NP_564573.3 | 20 - 21 | 2199 | 113 | ||
GenBank mRNA | AK228745.1 | - | 20 - 21 | 2054 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |