Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333450.1 | NP_001320603.1 | 5 - 6 | 1826 | 117 | |
NM_103952.4 | NP_175485.1 | 6 - 7 | 1843 | 117 | ||
GenBank mRNA | AK175623.1 | - | 6 - 7 | 1693 | 117 | |
BT012659.1 | - | 6 - 7 | 1393 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |