Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103994.2 | NP_175527.2 | 2 - 3 | 504 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |