Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121052.3 | NP_196576.1 | 6 - 7 | 601 | 65 | |
GenBank mRNA | AF116527.1 | - | 6 - 7 | 601 | 65 | |
AF537203.1 | - | 6 - 7 | 492 | 65 | ||
AY964092.1 | - | 6 - 7 | 492 | 65 | ||
AY964093.1 | - | 6 - 7 | 492 | 65 | ||
AY964094.1 | - | 6 - 7 | 524 | 65 | ||
AY964095.1 | - | 6 - 7 | 533 | 65 | ||
AY964097.1 | - | 6 - 7 | 492 | 65 | ||
BT030637.1 | - | 6 - 7 | 492 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Arabidopsis thaliana Probe spans exons |