Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKC;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121065.3 | NP_196589.1 | 7 - 8 | 727 | 102 | |
GenBank mRNA | AF375437.1 | - | 7 - 8 | 704 | 102 | |
AY120690.1 | - | 7 - 8 | 613 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |