Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121110.2 | NP_196633.2 | 10 - 11 | 2141 | 70 | |
GenBank mRNA | DQ167579.1 | - | 10 - 11 | 2141 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |