Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161232.2 | NP_001154704.2 | 4 - 5 | 585 | 73 | |
NM_001161233.1 | NP_001154705.1 | 4 - 5 | 544 | 73 | ||
NM_001343149.1 | NP_001332043.1 | 4 - 5 | 547 | 73 | ||
NM_001343150.1 | NP_001332042.1 | 4 - 5 | 547 | 73 | ||
NM_001343151.1 | NP_001332040.1 | 4 - 5 | 547 | 73 | ||
NM_001343152.1 | NP_001332041.1 | 4 - 5 | 544 | 73 | ||
NM_001343153.1 | NP_001332045.1 | 4 - 5 | 544 | 73 | ||
NM_001343154.1 | NP_001332044.1 | 4 - 5 | 544 | 73 | ||
NM_121141.5 | NP_196664.4 | 4 - 5 | 585 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |