Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121164.5 | NP_568246.1 | 3 - 4 | 705 | 75 | |
GenBank mRNA | AB005295.1 | - | 3 - 4 | 623 | 75 | |
AB005456.1 | - | 3 - 4 | 623 | 75 | ||
AK229362.1 | - | 3 - 4 | 585 | 75 | ||
BT025519.1 | - | 3 - 4 | 432 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |