Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085101.2 | NP_001078570.1 | 6 - 7 | 1322 | 71 | |
NM_001343183.1 | NP_001332634.1 | 6 - 7 | 1321 | 71 | ||
NM_001343184.1 | NP_001332633.1 | 6 - 7 | 1321 | 71 | ||
NM_121176.4 | NP_196699.1 | 6 - 7 | 1321 | 71 | ||
GenBank mRNA | AK227249.1 | - | 6 - 7 | 1315 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |