Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036794.2 | NP_001031871.1 | 14 - 15 | 1633 | 76 | |
NM_001036795.2 | NP_001031872.1 | 14 - 15 | 1504 | 76 | ||
NM_121275.4 | NP_568270.3 | 14 - 15 | 1505 | 76 | ||
GenBank mRNA | AK222187.1 | - | 14 - 15 | 1610 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |