Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036794.2 | NP_001031871.1 | 8 - 9 | 1017 | 93 | |
NM_001036795.2 | NP_001031872.1 | 8 - 9 | 888 | 93 | ||
NM_121275.4 | NP_568270.3 | 8 - 9 | 889 | 93 | ||
GenBank mRNA | AK222187.1 | - | 8 - 9 | 994 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |