Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DiT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125744.1 | NP_001119216.1 | 1 - 2 | 1120 | 66 | |
NM_121289.4 | NP_568283.2 | 1 - 2 | 1459 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |