Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128011.5 | NP_180026.1 | 3 - 4 | 951 | 83 | |
GenBank mRNA | AF095588.1 | - | 3 - 4 | 840 | 83 | |
AY062620.1 | - | 3 - 4 | 883 | 83 | ||
AY114695.1 | - | 3 - 4 | 822 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |