Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIC55-II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128041.4 | NP_180055.1 | 2 - 3 | 595 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |