Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128068.3 | NP_565585.1 | 1 - 2 | 410 | 107 | |
GenBank mRNA | AY057588.1 | - | 1 - 2 | 363 | 107 | |
AY143822.1 | - | 1 - 2 | 100 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |