Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335976.1 | NP_001324292.1 | 3 - 4 | 328 | 87 | |
NM_001335977.1 | NP_001324293.1 | 3 - 4 | 355 | 87 | ||
NM_001335978.1 | NP_001324291.1 | 2 - 3 | 277 | 87 | ||
NM_128074.4 | NP_565587.1 | 3 - 4 | 301 | 87 | ||
GenBank mRNA | AF185577.1 | - | 3 - 4 | 301 | 87 | |
AF185578.1 | - | 2 - 3 | 281 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |