Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128075.7 | NP_180090.6 | 1 - 2 | 445 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |