Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128127.4 | NP_180139.1 | 1 | 1388 | 92 | |
GenBank mRNA | AK221594.1 | - | 1 | 1335 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |