Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336027.1 | NP_001325078.1 | 5 - 6 | 1656 | 153 | |
NM_001336028.1 | NP_001325077.1 | 6 - 7 | 1512 | 153 | ||
NM_128148.4 | NP_180159.2 | 4 - 5 | 498 | 153 | ||
GenBank mRNA | AK175188.1 | - | 4 - 5 | 462 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |