Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CID7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336049.1 | NP_001324897.1 | 4 - 5 | 1611 | 72 | |
NM_001336050.1 | NP_001324896.1 | 4 - 5 | 1611 | 72 | ||
NM_001336052.1 | NP_001324892.1 | 4 - 5 | 1459 | 72 | ||
NM_001336054.1 | NP_001324894.1 | 4 - 5 | 1638 | 72 | ||
NM_128185.3 | NP_180196.1 | 4 - 5 | 1470 | 72 | ||
GenBank mRNA | BT030338.1 | - | 2 - 3 | 1212 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |