Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 11 - 12 | 1628 | 77 | |
GenBank mRNA | AY035110.1 | - | 11 - 12 | 937 | 77 | |
FJ708701.1 | - | 11 - 12 | 861 | 77 | ||
U47029.1 | - | 11 - 12 | 911 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |