Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 13 - 14 | 1775 | 73 | |
GenBank mRNA | AY035110.1 | - | 13 - 14 | 1084 | 73 | |
FJ708701.1 | - | 13 - 14 | 1008 | 73 | ||
U47029.1 | - | 13 - 14 | 1058 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |