Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 21 - 22 | 2343 | 151 | |
GenBank mRNA | AY035110.1 | - | 21 - 22 | 1652 | 151 | |
FJ708701.1 | - | 21 - 22 | 1576 | 151 | ||
U47029.1 | - | 21 - 22 | 1626 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |