Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084493.1 | NP_001077962.1 | 3 - 4 | 353 | 93 | |
NM_001084494.1 | NP_001077963.1 | 2 - 3 | 444 | 93 | ||
NM_001336070.1 | NP_001324176.1 | 3 - 4 | 360 | 93 | ||
NM_128200.5 | NP_565622.1 | 3 - 4 | 383 | 93 | ||
GenBank mRNA | AK226505.1 | - | 3 - 4 | 326 | 93 | |
AK317155.1 | - | 2 - 3 | 414 | 93 | ||
AY086218.1 | - | 3 - 4 | 354 | 93 | ||
AY150419.1 | - | 2 - 3 | 236 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |