Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT76D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128205.3 | NP_180216.1 | 1 - 2 | 535 | 81 | |
GenBank mRNA | KJ138896.1 | - | 1 - 2 | 461 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |